RIE156330261 Rieke Metals, Inc.

Ethyl 4-(3-fluorophenyl)-4-oxobutyrate

  • MDL number MFCD01320234
  • Cas Number : 898752-08-4
  • Molecular Weight : 224.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7542B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7542B
Availability details for
Quantity
  • Availability
  • Add to Cart