RIE156330262 Rieke Metals, Inc.

Ethyl 5-(3-fluorophenyl)-5-oxovalerate

  • MDL number MFCD01311616
  • Cas Number : 898752-10-8
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7542C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7542C
Availability details for
Quantity
  • Availability
  • Add to Cart