RIE156330263 Rieke Metals, Inc.

ethyl 6-(3-fluorophenyl)-6-oxohexanoate

  • MDL number MFCD01311615
  • Cas Number : 898752-12-0
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7542D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7542D
Availability details for
Quantity
  • Availability
  • Add to Cart