RIE156330264 Rieke Metals, Inc.

ethyl 7-(3-fluorophenyl)-7-oxoheptanoate

  • MDL number MFCD01311614
  • Cas Number : 122115-57-5
  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7542E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7542E
Availability details for
Quantity
  • Availability
  • Add to Cart