RIE156330266 Rieke Metals, Inc.

Ethyl 4-(3-chlorophenyl)-4-oxobutyrate

  • MDL number MFCD01320236
  • Cas Number : 147374-00-3
  • Molecular Weight : 240.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7544B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7544B
Availability details for
Quantity
  • Availability
  • Add to Cart