RIE156330276 Rieke Metals, Inc.

Ethyl 5-(3,5-difluorophenyl)-5-oxovalerate

  • MDL number MFCD01320247
  • Cas Number : 898752-37-9
  • Molecular Weight : 256.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7548C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7548C
Availability details for
Quantity
  • Availability
  • Add to Cart