RIE156330277 Rieke Metals, Inc.

ethyl 6-(3,5-difluorophenyl)-6-oxohexanoate

  • MDL number MFCD01320248
  • Cas Number : 898752-40-4
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7548D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7548D
Availability details for
Quantity
  • Availability
  • Add to Cart