RIE156330279 Rieke Metals, Inc.

ethyl 8-(3,5-difluorophenyl)-8-oxooctanoate

  • MDL number MFCD01320250
  • Cas Number : 898752-46-0
  • Molecular Weight : 298.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7548F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7548F
Availability details for
Quantity
  • Availability
  • Add to Cart