RIE156330280 Rieke Metals, Inc.

ethyl 4-oxo-4-(3,4,5-trifluorophenyl)butyrate

  • MDL number MFCD01320251
  • Cas Number : 898752-49-3
  • Molecular Weight : 260.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7550B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7550B
Availability details for
Quantity
  • Availability
  • Add to Cart