RIE156330281 Rieke Metals, Inc.

Ethyl 5-oxo-5-(3,4,5-trifluorophenyl)valerate

  • MDL number MFCD01320252
  • Cas Number : 898752-52-8
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7550C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7550C
Availability details for
Quantity
  • Availability
  • Add to Cart