RIE156330284 Rieke Metals, Inc.

ethyl 4-(4-fluoro-3-methylphenyl)-4-oxobutyrate

  • MDL number MFCD07700153
  • Cas Number : 898752-61-9
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7554B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7554B
Availability details for
Quantity
  • Availability
  • Add to Cart