RIE156330286 Rieke Metals, Inc.

ethyl 4-oxo-4-(4-phenoxyphenyl)butyrate

  • MDL number MFCD01320257
  • Cas Number : 52888-63-8
  • Molecular Weight : 298.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7555B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7555B
Availability details for
Quantity
  • Availability
  • Add to Cart