RIE156330290 Rieke Metals, Inc.

Ethyl 7-oxo-7-(3-phenoxyphenyl)heptanoate

  • MDL number MFCD02261451
  • Cas Number : 898752-70-0
  • Molecular Weight : 340.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7556E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7556E
Availability details for
Quantity
  • Availability
  • Add to Cart