RIE156330291 Rieke Metals, Inc.

ethyl 4-oxo-4-(4-thiomethylphenyl)butyrate

  • MDL number MFCD01320259
  • Cas Number : 7028-70-8
  • Molecular Weight : 252.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7557B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7557B
Availability details for
Quantity
  • Availability
  • Add to Cart