RIE156330294 Rieke Metals, Inc.

ethyl 7-(2-methoxyphenyl)-7-oxoheptanoate

  • MDL number MFCD01311630
  • Cas Number : 122115-56-4
  • Molecular Weight : 278.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7560E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7560E
Availability details for
Quantity
  • Availability
  • Add to Cart