RIE156330295 Rieke Metals, Inc.

Ethyl 5-(1-naphthyl)-5-oxovalerate

  • MDL number MFCD01320263
  • Cas Number : 40335-93-1
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7561C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7561C
Availability details for
Quantity
  • Availability
  • Add to Cart