RIE156330296 Rieke Metals, Inc.

ethyl 6-(1-naphthyl)-6-oxohexanoate

  • MDL number MFCD01320264
  • Cas Number : 101743-65-1
  • Molecular Weight : 284.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7561D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7561D
Availability details for
Quantity
  • Availability
  • Add to Cart