RIE156330299 Rieke Metals, Inc.

Ethyl 5-(2-naphthyl)-5-oxovalerate

  • MDL number MFCD01320267
  • Cas Number : 109089-73-8
  • Molecular Weight : 270.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7562C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7562C
Availability details for
Quantity
  • Availability
  • Add to Cart