RIE156330301 Rieke Metals, Inc.

Ethyl 4-oxo-4-(9-Phenanthryl)butyrate

  • MDL number MFCD01320270
  • Cas Number : 898752-85-7
  • Molecular Weight : 306.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7563B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7563B
Availability details for
Quantity
  • Availability
  • Add to Cart