RIE156330302 Rieke Metals, Inc.

Ethyl 5-oxo-5-(9-Phenanthryl)valerate

  • MDL number MFCD01320271
  • Cas Number : 898752-88-0
  • Molecular Weight : 320.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7563C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7563C
Availability details for
Quantity
  • Availability
  • Add to Cart