RIE156330305 Rieke Metals, Inc.

Ethyl 8-oxo-8-(9-Phenanthryl)octanoate

  • MDL number MFCD02261638
  • Cas Number : 898752-94-8
  • Molecular Weight : 362.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7563F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7563F
Availability details for
Quantity
  • Availability
  • Add to Cart