RIE156330308 Rieke Metals, Inc.

ethyl 6-(2,3-difluorophenyl)-6-oxohexanoate

  • MDL number MFCD01320279
  • Cas Number : 898753-00-9
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7565D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7565D
Availability details for
Quantity
  • Availability
  • Add to Cart