RIE156330311 Rieke Metals, Inc.

ethyl 4-(2,4-difluorophenyl)-4-oxobutyrate

  • MDL number MFCD01320281
  • Cas Number : 898753-06-5
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7566B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7566B
Availability details for
Quantity
  • Availability
  • Add to Cart