RIE156330317 Rieke Metals, Inc.

Ethyl 6-(2,5-difluorophenyl)-6-oxohexanoate

  • MDL number MFCD01320287
  • Cas Number : 898753-18-9
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7567D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7567D
Availability details for
Quantity
  • Availability
  • Add to Cart