RIE156330320 Rieke Metals, Inc.

Ethyl 4-(2,6-difluorophenyl)-4-oxobutyrate

  • MDL number MFCD01320289
  • Cas Number : 493004-53-8
  • Molecular Weight : 242.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7568B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7568B
Availability details for
Quantity
  • Availability
  • Add to Cart