RIE156330323 Rieke Metals, Inc.

ethyl 7-(2,6-difluorophenyl)-7-oxoheptanoate

  • MDL number MFCD01311651
  • Cas Number : 898753-28-1
  • Molecular Weight : 284.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7568E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7568E
Availability details for
Quantity
  • Availability
  • Add to Cart