RIE156330326 Rieke Metals, Inc.

Ethyl 5-(2-fluorophenyl)-5-oxovalerate

  • MDL number MFCD01320294
  • Cas Number : 898753-35-0
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7569C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7569C
Availability details for
Quantity
  • Availability
  • Add to Cart