RIE156330327 Rieke Metals, Inc.

ethyl 6-(2-fluorophenyl)-6-oxohexanoate

  • MDL number MFCD01320295
  • Cas Number : 898753-38-3
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7569D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7569D
Availability details for
Quantity
  • Availability
  • Add to Cart