RIE156330328 Rieke Metals, Inc.

ethyl 7-(2-fluorophenyl)-7-oxoheptanoate

  • MDL number MFCD01311652
  • Cas Number : 898753-41-8
  • Molecular Weight : 266.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7569E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7569E
Availability details for
Quantity
  • Availability
  • Add to Cart