RIE156330330 Rieke Metals, Inc.

Ethyl 4-(3-cyanophenyl)-4-oxobutyrate

  • MDL number MFCD01320297
  • Molecular Weight : 231.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7570B

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7570B
Availability details for
Quantity
  • Availability
  • Add to Cart