RIE156330331 Rieke Metals, Inc.

Ethyl-5-(3-cyanophenyl)-5-oxovalerate

  • MDL number MFCD01320298
  • Cas Number : 898753-47-4
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7570C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7570C
Availability details for
Quantity
  • Availability
  • Add to Cart