RIE156330332 Rieke Metals, Inc.

Ethyl 6-(3-cyanophenyl)-6-oxohexanoate

  • MDL number MFCD01320299
  • Molecular Weight : 259.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7570D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7570D
Availability details for
Quantity
  • Availability
  • Add to Cart