RIE156330334 Rieke Metals, Inc.

Ethyl 8-(3-cyanophenyl)-8-oxooctanoate

  • MDL number MFCD01320300
  • Molecular Weight : 287.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7570F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7570F
Availability details for
Quantity
  • Availability
  • Add to Cart