RIE156330337 Rieke Metals, Inc.

Ethyl 7-(4-cyanophenyl)-7-oxoheptanoate

  • MDL number MFCD01311656
  • Molecular Weight : 273.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7571E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7571E
Availability details for
Quantity
  • Availability
  • Add to Cart