RIE156330342 Rieke Metals, Inc.

ethyl 7-(4-biphenyl)-7-oxoheptanoate

  • MDL number MFCD01311658
  • Cas Number : 147862-41-7
  • Molecular Weight : 324.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7572E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7572E
Availability details for
Quantity
  • Availability
  • Add to Cart