RIE156330343 Rieke Metals, Inc.

ethyl 2,2-dimethyl-4-(3-methoxyphenyl)-4-oxobutyrate

  • MDL number MFCD01320309
  • Cas Number : 898753-53-2
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7573

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7573
Availability details for
Quantity
  • Availability
  • Add to Cart