RIE156330344 Rieke Metals, Inc.

Ethyl 2,2-dimethyl-4-(4-methoxyphenyl)-4-oxobutyrate

  • MDL number MFCD01320310
  • Cas Number : 898753-56-5
  • Molecular Weight : 264.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7574

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7574
Availability details for
Quantity
  • Availability
  • Add to Cart