RIE156330345 Rieke Metals, Inc.

ethyl 4-(3,4-difluorophenyl)-2,2-dimethyl-4-oxobutyrate

  • MDL number MFCD01320312
  • Cas Number : 898753-59-8
  • Molecular Weight : 270.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7576

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7576
Availability details for
Quantity
  • Availability
  • Add to Cart