RIE156330347 Rieke Metals, Inc.

ethyl 2,2-dimethyl-4-(3-fluorophenyl)-4-oxobutyrate

  • MDL number MFCD01320314
  • Cas Number : 898753-65-6
  • Molecular Weight : 252.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7578

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7578
Availability details for
Quantity
  • Availability
  • Add to Cart