RIE156330350 Rieke Metals, Inc.

ethyl 2,2-dimethyl-4-oxo-4-(4-thiomethylphenyl)butyrate

  • MDL number MFCD01320317
  • Cas Number : 898753-74-7
  • Molecular Weight : 280.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7581

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7581
Availability details for
Quantity
  • Availability
  • Add to Cart