RIE156330351 Rieke Metals, Inc.

Ethyl 5-cyclohexyl-5-oxovalerate

  • MDL number MFCD01320319
  • Cas Number : 16076-61-2
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7582C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7582C
Availability details for
Quantity
  • Availability
  • Add to Cart