RIE156330352 Rieke Metals, Inc.

ethyl 7-cyclohexyl-7-oxoheptanoate

  • MDL number MFCD01311660
  • Cas Number : 898753-77-0
  • Molecular Weight : 254.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7582E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7582E
Availability details for
Quantity
  • Availability
  • Add to Cart