RIE156330354 Rieke Metals, Inc.

Ethyl 5-cyclopentyl-5-oxovalerate

  • MDL number MFCD01320323
  • Cas Number : 24071-97-4
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7583C

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7583C
Availability details for
Quantity
  • Availability
  • Add to Cart