RIE156330355 Rieke Metals, Inc.

ethyl 6-cyclopentyl-6-oxohexanoate

  • MDL number MFCD01320324
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7583D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7583D
Availability details for
Quantity
  • Availability
  • Add to Cart