RIE156330360 Rieke Metals, Inc.

ethyl 6-cyclobutyl-6-oxohexanoate

  • MDL number MFCD01320328
  • Cas Number : 898776-18-6
  • Molecular Weight : 212.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7584D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7584D
Availability details for
Quantity
  • Availability
  • Add to Cart