RIE156330361 Rieke Metals, Inc.

ethyl 7-cyclobutyl-7-oxoheptanoate

  • MDL number MFCD01311664
  • Cas Number : 898776-21-1
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7584E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7584E
Availability details for
Quantity
  • Availability
  • Add to Cart