RIE156330366 Rieke Metals, Inc.

ethyl 8-cyclopropyl-8-oxooctanoate

  • MDL number MFCD01320333
  • Cas Number : 898776-36-8
  • Molecular Weight : 226.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7585F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7585F
Availability details for
Quantity
  • Availability
  • Add to Cart