RIE156330370 Rieke Metals, Inc.

ethyl 8,8-dimethyl-7-oxononanoate

  • MDL number MFCD01311669
  • Cas Number : 898776-42-6
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7586E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7586E
Availability details for
Quantity
  • Availability
  • Add to Cart