RIE156330371 Rieke Metals, Inc.

Ethyl 9,9-dimethyl-8-oxodecanoate

  • MDL number MFCD01320337
  • Cas Number : 898776-45-9
  • Molecular Weight : 242.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7586F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7586F
Availability details for
Quantity
  • Availability
  • Add to Cart