RIE156330375 Rieke Metals, Inc.

ethyl 6-oxoundecanoate

  • MDL number MFCD01320345
  • Cas Number : 72665-99-7
  • Molecular Weight : 228.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/7589D

Safety Information

MSDS http://riekemetals.com/products-services/catnum/7589D
Availability details for
Quantity
  • Availability
  • Add to Cart